Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mprip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012027.2 | NP_036157.2 | 8 - 9 | 1370 | 92 | |
NM_201245.3 | NP_957697.1 | 8 - 9 | 1370 | 92 | ||
GenBank mRNA | AK035435.1 | - | 2 - 3 | 660 | 92 | |
AK154807.1 | - | 9 - 10 | 1377 | 92 | ||
AK155630.1 | - | 8 - 9 | 1151 | 92 | ||
AK158296.1 | - | 8 - 9 | 1359 | 92 | ||
AK162530.1 | - | 5 - 6 | 701 | 92 | ||
AK170849.1 | - | 8 - 9 | 1354 | 92 | ||
AK171089.1 | - | 8 - 9 | 1359 | 92 | ||
BC039932.1 | - | 7 - 8 | 894 | 92 | ||
BC049803.1 | - | 8 - 9 | 896 | 92 | ||
BC063067.1 | - | 8 - 9 | 1333 | 5721396 | 92 | |
U73200.1 | - | 8 - 9 | 1230 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |