Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmynd15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029929.2 | NP_001025100.1 | 5 - 6 | 1598 | 86 | |
GenBank mRNA | AK029988.1 | - | 5 - 6 | 1599 | 86 | |
AK128979.1 | - | 4 - 5 | 1905 | 86 | ||
AK171931.1 | - | 5 - 6 | 1447 | 86 | ||
BC038060.1 | - | 2 - 3 | 208 | 86 | ||
BC151086.1 | - | 5 - 6 | 1270 | 9087999 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |