Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900001946.mm.1 |
NM_001129787.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001946.mm.1 |
NM_001129787.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001946.mm.1 |
TC0900000259.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900001947.mm.1 |
NM_001129787.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001947.mm.1 |
NM_001129787.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001947.mm.1 |
TC0900000259.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900001949.mm.1 |
NM_001129787.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001949.mm.1 |
NM_001129787.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001949.mm.1 |
TC0900000259.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900001950.mm.1 |
NM_001129787.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001950.mm.1 |
NM_001129787.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001950.mm.1 |
TC0900000259.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900001948.mm.1 |
NM_001129787.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001948.mm.1 |
NM_001129787.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001948.mm.1 |
TC0900000259.mm.1 |
MTA TC |