Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nutm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033400.2 | NP_001028572.1 | 2 - 3 | 842 | 53 | |
GenBank mRNA | AK133253.1 | - | 2 - 3 | 843 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |