Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1964325 |
BC055379.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1964325 |
BC100580.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1964325 |
AK030702.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1964325 |
NM_030599.4 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2106681 |
M77677.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2106681 |
BC055379.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2106681 |
BC100580.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2106681 |
AK030702.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2106681 |
NM_030599.4 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P109570 |
BC055379.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P109570 |
BC100580.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P109570 |
AK030702.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P109570 |
AK138766.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P109570 |
NM_030599.4 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
851935 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
425160 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
715631 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
868290 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
821853 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
65008 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
100020 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
588876 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
315103 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
27189 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
209483 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
415253 |
AK048522.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
415253 |
AK158395.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
415253 |
NM_177709.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
506005 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
779155 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
668287 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
111140 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
164492 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
853000 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
478873 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
242593 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
81837 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
44875 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
167076 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P178521 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2106681 |
M77677.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2106681 |
BC055379.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2106681 |
BC100580.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2106681 |
AK030702.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2106681 |
NM_030599.4 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1964325 |
BC055379.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1964325 |
BC100580.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1964325 |
AK030702.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1964325 |
NM_030599.4 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P109570 |
BC055379.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P109570 |
BC100580.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P109570 |
AK030702.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P109570 |
AK138766.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P109570 |
NM_030599.4 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24840978 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24840976 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24840979 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24840980 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24840981 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24840986 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24840984 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24840985 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06066652 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P09512880 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P01444938 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P178521 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P09512909 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P122810 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P178521 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P09512880 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P09512909 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P01444938 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24841002 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24840994 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24840992 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24840978 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24840989 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24840984 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24840999 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24840981 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24840985 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24840980 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24840980 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05191366 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24840988 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05819214 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24840978 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24840994 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24840985 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24840984 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05484229 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24840992 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05130649 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05167053 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24840979 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24840989 |
- |
- |