Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300006461.mm.1 |
NM_001013804.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300006461.mm.1 |
NM_001013804.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300006461.mm.1 |
TC0300000843.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300006463.mm.1 |
NM_001013804.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300006463.mm.1 |
NM_001013804.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300006463.mm.1 |
TC0300000843.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300006467.mm.1 |
NM_001013804.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300006467.mm.1 |
TC0300000843.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300006464.mm.1 |
NM_001013804.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300006464.mm.1 |
NM_001013804.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300006464.mm.1 |
TC0300000843.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300006462.mm.1 |
NM_001013804.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300006462.mm.1 |
NM_001013804.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300006462.mm.1 |
TC0300000843.mm.1 |
MTA TC |