Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039047.1 | NP_001034136.1 | 2 - 3 | 521 | 69 | |
GenBank mRNA | BC147805.1 | - | 2 - 3 | 540 | 9087522 | 69 |
BC147806.1 | - | 2 - 3 | 540 | 9087523 | 69 | |
BC150936.1 | - | 2 - 3 | 595 | 9087847 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |