Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgfr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163215.2 | NP_001156687.1 | 9 | 1478 | 151 | |
NM_001163216.2 | NP_001156688.1 | 7 | 1193 | 151 | ||
NM_001163217.2 | NP_001156689.1 | 8 | 1253 | 151 | ||
NM_001205270.1 | NP_001192199.1 | 9 | 1452 | 151 | ||
NM_008010.5 | NP_032036.2 | 9 | 1452 | 151 | ||
GenBank mRNA | AF024638.1 | - | 7 | 1101 | 151 | |
AK143592.1 | - | 9 | 1385 | 151 | ||
AK163495.1 | - | 7 | 1031 | 151 | ||
BC053056.1 | - | 9 | 1362 | 5708838 | 151 | |
BU053488.1 | - | 4 | 468 | 151 | ||
M81342.1 | - | 9 | 1370 | 151 | ||
S56291.1 | - | 8 | 1197 | 151 | ||
X58255.1 | - | 9 | 1356 | 151 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Both primers and probe map within a single exon |