Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010660.2 | NP_034790.2 | 5 - 6 | 1152 | 66 | |
GenBank mRNA | AK014360.1 | - | 5 - 6 | 1138 | 66 | |
AK076508.1 | - | 5 - 6 | 1150 | 66 | ||
AK081402.1 | - | 5 - 6 | 1152 | 66 | ||
AK081914.1 | - | 5 - 6 | 1149 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |