Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2g6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199024.1 | NP_001185953.1 | 8 - 9 | 1962 | 61 | |
NM_001199025.1 | NP_001185954.1 | 9 - 10 | 1467 | 61 | ||
NM_016915.4 | NP_058611.1 | 8 - 9 | 1508 | 61 | ||
GenBank mRNA | AK144495.1 | - | 9 - 10 | 1341 | 61 | |
AK153333.1 | - | 8 - 9 | 1768 | 61 | ||
BC003487.1 | - | 8 - 9 | 1301 | 2811357 | 61 | |
BC049778.1 | - | 8 - 9 | 1688 | 61 | ||
BC052845.1 | - | 8 - 9 | 1333 | 30054573 | 61 | |
U88624.2 | - | 7 - 8 | 1184 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |