Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK165594.1 | - | 1313 | 86 | ||
AK168270.1 | - | 9 - 10 | 1259 | 86 | ||
BC003799.1 | - | 9 - 10 | 1275 | 3488471 | 86 | |
U10119.1 | - | 9 - 10 | 1319 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |