Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600002393.mm.1 |
TC0600000325.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600002393.mm.1 |
NM_172398.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002393.mm.1 |
NM_172398.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002405.mm.1 |
TC0600000325.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600002405.mm.1 |
NM_172398.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002405.mm.1 |
NM_172398.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002401.mm.1 |
TC0600000325.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600002401.mm.1 |
NM_172398.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002401.mm.1 |
NM_172398.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002403.mm.1 |
TC0600000325.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600002403.mm.1 |
NM_172398.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002403.mm.1 |
NM_172398.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002391.mm.1 |
TC0600000325.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600002391.mm.1 |
NM_172398.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002391.mm.1 |
NM_172398.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002404.mm.1 |
TC0600000325.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600002404.mm.1 |
NM_172398.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002404.mm.1 |
NM_172398.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002388.mm.1 |
TC0600000325.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600002388.mm.1 |
NM_172398.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002388.mm.1 |
NM_172398.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002390.mm.1 |
TC0600000325.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600002390.mm.1 |
NM_172398.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002390.mm.1 |
NM_172398.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002397.mm.1 |
TC0600000325.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600002397.mm.1 |
NM_172398.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002397.mm.1 |
NM_172398.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002398.mm.1 |
TC0600000325.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600002398.mm.1 |
NM_172398.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002398.mm.1 |
NM_172398.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002402.mm.1 |
TC0600000325.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600002402.mm.1 |
NM_172398.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002402.mm.1 |
NM_172398.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002392.mm.1 |
TC0600000325.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600002392.mm.1 |
NM_172398.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002392.mm.1 |
NM_172398.3 |
Refseq |