Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284411.1 | NP_001271340.1 | 5 - 6 | 491 | 141 | |
NM_026776.4 | NP_081052.2 | 5 - 6 | 467 | 141 | ||
GenBank mRNA | AK002500.1 | - | 5 - 6 | 429 | 141 | |
AK003715.2 | - | 5 - 6 | 439 | 141 | ||
AK005149.1 | - | 5 - 6 | 421 | 140 | ||
AK145811.1 | - | 5 - 6 | 438 | 141 | ||
AK150822.1 | - | 5 - 6 | 449 | 141 | ||
AK166008.1 | - | 5 - 6 | 446 | 141 | ||
AK212846.1 | - | 2 - 3 | 116 | 141 | ||
BC029181.1 | - | 5 - 6 | 431 | 4240500 | 141 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |