Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdha5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009959.1 | NP_034089.1 | 1 | 649 | 90 | |
GenBank mRNA | AB008181.1 | - | 1 | 542 | 90 | |
AY013763.1 | - | 1 | 649 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |