Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmn2r56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104648.1 | NP_001098118.1 | 3 - 4 | 1278 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Recommended Assay Mus musculus Probe spans exons |