Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgcz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145841.2 | NP_665840.2 | 7 - 8 | 1479 | 77 | |
GenBank mRNA | AK041231.1 | - | 7 - 8 | 1084 | 77 | |
AK048960.1 | - | 7 - 8 | 1443 | 77 | ||
AK136779.1 | - | 7 - 8 | 1480 | 77 | ||
AY095374.1 | - | 6 - 7 | 695 | 77 | ||
AY250713.1 | - | 6 - 7 | 695 | 77 | ||
BC104048.1 | - | 6 - 7 | 695 | 40040143 | 77 | |
BC104049.2 | - | 6 - 7 | 695 | 40040144 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons |