Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlx6os2 | |||||||
---|---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_002839.2 | 1 | 960 | 75 | |||
GenBank mRNA | AF022079.1 | - | LIT3325 Dlx6as |
1 | 957 | 75 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | split hand/foot malformation (ectrodactyly) type 1 | Shfm1 |
DSS1, Shfdg1, Shfg |
-285.1kb | Sense | Chr. 6 - 6558275 - 6578658 on NCBI Build GRCm38 |
Downstream | distal-less homeobox 5 | Dlx5 | AI385752 | 12.7kb | Sense | Chr. 6 - 6877801 - 6882068 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |