Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007554.2 | NP_031580.2 | 1 | 156 | 97 | |
GenBank mRNA | AK136527.1 | - | 1 | 157 | 97 | |
AK145359.1 | - | 1 | 226 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |