Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix HT MG-430 PM Array Plate |
1438273_PM_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0200009701.mm.1 |
NM_001145100.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200009701.mm.1 |
NM_001145100.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200009701.mm.1 |
TC0200001217.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200009698.mm.1 |
NM_001145100.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200009698.mm.1 |
NM_001145100.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200009698.mm.1 |
TC0200001217.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200009700.mm.1 |
NM_001145100.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200009700.mm.1 |
NM_001145100.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200009700.mm.1 |
TC0200001217.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200009704.mm.1 |
NM_001145100.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200009704.mm.1 |
NM_001145100.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200009704.mm.1 |
TC0200001217.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200009703.mm.1 |
NM_001145100.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200009703.mm.1 |
NM_001145100.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200009703.mm.1 |
TC0200001217.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200009699.mm.1 |
NM_001145100.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200009699.mm.1 |
NM_001145100.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200009699.mm.1 |
TC0200001217.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200009705.mm.1 |
NM_001145100.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200009705.mm.1 |
NM_001145100.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200009705.mm.1 |
TC0200001217.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200009702.mm.1 |
NM_001145100.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200009702.mm.1 |
NM_001145100.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200009702.mm.1 |
TC0200001217.mm.1 |
MTA TC |
Mouse Expression Set 430 |
1438273_at |
- |
- |
Mouse Expression Set 430 |
1438273_PM_at |
- |
- |