Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A330087I24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK039689.1 | - | 6 - 7 | 1577 | 71 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
vomeronasal 2, receptor 84 |
Vmn2r84 | EG625068 | Sense | Chr. 10 - 130385800 - 130394241 on NCBI Build GRCm38 | |
Downstream |
vomeronasal 2, receptor 85 |
Vmn2r85 | EG623734 | 30.3kb | Sense | Chr. 10 - 130418261 - 130429612 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |