Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | estrogen-related receptor gamma | Esrrg |
ERR3, Errg, NR3B3, mKIAA0832 |
-368.2kb | Anti-sense | Chr. 1 - 187608792 - 188214884 on NCBI Build GRCm38 |
Downstream | potassium channel tetramerisation domain containing 3 | Kctd3 |
4930438A20Rik, 9330185B06, E330032J19Rik, NY-REN-45 |
377.9kb | Sense | Chr. 1 - 188971095 - 189007868 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |