Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK140442.1 | - | 1 | 753 | 104 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | lysine (K)-specific methyltransferase 2A | Kmt2a |
6430520K01, ALL-1, All1, Cxxc7, HRX, HTRX1, Mll, Mll1, mKIAA4050 |
-34.4kb | Sense | Chr. 9 - 44803355 - 44881359 on NCBI Build GRCm38 |
Downstream | ubiquitination factor E4A | Ube4a |
4732444G18Rik, 9930123J21Rik, UFD2b |
4.3kb | Sense | Chr. 9 - 44923127 - 44965600 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |