Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC031980.1 | - | 1 | 1950 | 75 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | beta-site APP-cleaving enzyme 2 | Bace2 |
1110059C24Rik, AEPLC, AI850424, ALP56, ARP1, ASP21, BAE2, CDA13, CEAP1, DRAP |
-19.8kb | Anti-sense | Chr. 16 - 97356728 - 97439012 on NCBI Build GRCm38 |
Downstream |
family with sequence similarity 3, member B |
Fam3b |
2-21, 9030624C24Rik, D16Jhu19e, ORF9, Pander |
10.3kb | Sense | Chr. 16 - 97471086 - 97504936 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |