Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prdm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256380.1 | NP_001243309.1 | 8 | 8325 | 94 | |
GenBank mRNA | AK014782.1 | - | 1 | 2054 | 94 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
kazrin, periplakin interacting protein |
Kazn |
2310007B04Rik, 9030409G11Rik, AI256607, AI666764, AW061099, Kaz, Kazrin |
-889.0kb | Sense | Chr. 4 - 142102390 - 142239401 on NCBI Build GRCm38 |
Downstream | microRNA 7021 | Mir7021 | mmu-mir-7021 | 5.5kb | Sense | Chr. 4 - 143136100 - 143136162 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |