Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2rx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK019679.1 | - | 1 - 2 | 194 | 104 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | proteoglycan 3 | Prg3 |
MBP2, MBPH |
-33.7kb | Anti-sense | Chr. 2 - 84988196 - 84996559 on NCBI Build GRCm38 |
Downstream | structure specific recognition protein 1 | Ssrp1 |
C81323, Hmg1-rs1, Hmgi-rs3, Hmgox, T160 |
3.9kb | Anti-sense | Chr. 2 - 85037120 - 85047114 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus 1700 Probe spans exons |