Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm17252 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK033745.1 | - | 1 - 2 | 72 | 98 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
predicted gene, 17365 |
Gm17365 |
Pate-dj, Pate3 |
-37.2kb | Sense | Chr. 9 - 35645113 - 35648202 on NCBI Build GRCm38 |
Downstream |
predicted gene, 17677 |
Gm17677 | Pate-n | 53.7kb | Anti-sense | Chr. 9 - 35741049 - 35742252 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |