Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | proline rich Gla (G-carboxyglutamic acid) 1 | Prrg1 | 2010007L08Rik | Sense | Chr. X - 78449610 - 78583913 on NCBI Build GRCm38 | |
Downstream |
family with sequence similarity 47, member C |
Fam47c |
4921509A18Rik, Fam47a, Fam47a-ps |
176.0kb | Anti-sense | Chr. X - 78737763 - 78739410 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |