Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scn9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK051636.1 | - | 1 | 1745 | 108 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | predicted gene 13629 | Gm13629 | OTTMUSG00000013098 | Anti-sense | Chr. 2 - 66440880 - 66547472 on NCBI Build GRCm38 | |
Downstream |
sodium channel, voltage-gated, type VII, alpha |
Scn7a |
1110034K09Rik, AI642000, NaG, Nav2, Nav2.3, Nax, Scn6a |
150.9kb | Sense | Chr. 2 - 66673424 - 66785091 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |