Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | protein O-linked mannose beta 1,4-N-acetylglucosaminyltransferase 2 | Pomgnt2 |
Ago61, C85492, Gtdc2 |
-13.8kb | Sense | Chr. 9 - 121981606 - 121996026 on NCBI Build GRCm38 |
Downstream | SNF related kinase | Snrk |
2010012F07Rik, AI448042, AW547029, E030034B15, R74830, mKIAA0096 |
105.1kb | Anti-sense | Chr. 9 - 122117266 - 122169707 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |