Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | paired related homeobox 1 | Prrx1 |
A230024N07Rik, AA755424, AI385634, AI843499, K-2, Pmx, Pmx1, Prx1, mHox |
Sense | Chr. 1 - 163245119 - 163313705 on NCBI Build GRCm38 | |
Downstream |
golgin, RAB6-interacting |
Gorab |
AI467484, NTKL-BP1, Scyl1bp1 |
82.5kb | Sense | Chr. 1 - 163384906 - 163403908 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |