Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cers3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164201.1 | NP_001157673.1 | 9 - 10 | 1095 | 75 | |
GenBank mRNA | DQ358087.1 | - | 8 - 9 | 853 | 75 | |
DQ646881.1 | - | 9 - 10 | 974 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |