Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsun6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001165941.1 | NP_001159413.1 | 3 - 4 | 657 | 67 | |
NM_001165942.1 | NP_001159414.1 | 2 - 3 | 544 | 67 | ||
NM_001165943.1 | NP_001159415.1 | 2 - 3 | 361 | 67 | ||
NM_028950.4 | NP_083226.3 | 2 - 3 | 544 | 67 | ||
GenBank mRNA | AK016629.1 | - | 2 - 3 | 362 | 67 | |
AK016811.1 | - | 3 - 4 | 670 | 67 | ||
AK042750.1 | - | 2 - 3 | 544 | 67 | ||
AK077778.1 | - | 2 - 3 | 428 | 67 | ||
AK132428.1 | - | 2 - 3 | 511 | 67 | ||
BC053751.1 | - | 2 - 3 | 512 | 6335896 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |