Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam160b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194345.1 | NP_919326.1 | 14 - 15 | 1906 | 85 | |
GenBank mRNA | AK090035.1 | - | 9 - 10 | 1157 | 85 | |
BC037228.1 | - | 2 - 3 | 183 | 85 | ||
BC050861.1 | - | 13 - 14 | 2029 | 85 | ||
BC058108.1 | - | 2 - 3 | 211 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |