Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300015727.mm.1 |
NM_001104615.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015727.mm.1 |
NM_001104615.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015727.mm.1 |
TC0300002083.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300015765.mm.1 |
NM_175674.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015765.mm.1 |
NM_175674.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015765.mm.1 |
TC0300002089.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300015746.mm.1 |
NM_001104619.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015746.mm.1 |
NM_001104619.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015746.mm.1 |
TC0300002087.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300015747.mm.1 |
NM_001104619.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015747.mm.1 |
NM_001104619.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015747.mm.1 |
TC0300002087.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300015749.mm.1 |
NM_001104619.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015749.mm.1 |
NM_001104619.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015749.mm.1 |
TC0300002087.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300015766.mm.1 |
NM_175674.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015766.mm.1 |
NM_175674.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015766.mm.1 |
TC0300002089.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300015728.mm.1 |
NM_001104615.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015728.mm.1 |
NM_001104615.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015728.mm.1 |
TC0300002083.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300015748.mm.1 |
NM_001104619.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015748.mm.1 |
NM_001104619.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015748.mm.1 |
TC0300002087.mm.1 |
MTA TC |