Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbed6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166552.1 | NP_001160024.1 | 1 | 3145 | 93 | |
GenBank mRNA | AK090300.1 | - | 1 | 737 | 93 | |
AK144981.1 | - | 1 | 3145 | 93 | ||
FM882123.1 | - | 1 | 1857 | 93 | ||
FR682664.1 | - | 3578 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon |