Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm17677 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167588.1 | NP_001161060.1 | 2 - 3 | 197 | 80 | |
GenBank mRNA | EU703628.1 | - | 2 - 3 | 197 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons |