Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ruvbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019685.2 | NP_062659.1 | 7 - 8 | 919 | 97 | |
GenBank mRNA | AF100694.1 | - | 7 - 8 | 958 | 97 | |
AK012170.1 | - | 7 - 8 | 864 | 97 | ||
AK132783.1 | - | 7 - 8 | 901 | 97 | ||
AK145062.1 | - | 868 | 97 | |||
AK146258.1 | - | 7 - 8 | 911 | 97 | ||
AK146374.1 | - | 7 - 8 | 912 | 97 | ||
AK156078.1 | - | 7 - 8 | 889 | 97 | ||
AK186720.1 | - | 1 - 2 | 70 | 97 | ||
BC004718.1 | - | 7 - 8 | 885 | 3582426 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |