Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkd1l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039700.2 | NP_001034789.2 | 9 - 10 | 2432 | 99 | |
NM_001286454.1 | NP_001273383.1 | 8 - 9 | 2402 | 99 | ||
NM_181544.2 | NP_853522.2 | 8 - 9 | 2402 | 99 | ||
GenBank mRNA | AB290926.1 | - | 8 - 9 | 2402 | 99 | |
AY164486.1 | - | 8 - 9 | 2402 | 99 | ||
DQ382344.1 | - | 8 - 9 | 2402 | 99 | ||
DQ382345.1 | - | 9 - 10 | 2432 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Detect Genomic DNA Mus musculus Probe spans exons |