Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.0 ST Array |
10526723 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10526719 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10526721 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10526718 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10526720 |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0500014987.mm.1 |
TC0500003715.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500014987.mm.1 |
NM_001039889.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500014980.mm.1 |
TC0500003714.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500014980.mm.1 |
NM_001126045.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500014980.mm.1 |
NM_001126045.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500014980.mm.1 |
NM_001168602.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500014980.mm.1 |
NM_001168602.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500014981.mm.1 |
TC0500003714.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500014981.mm.1 |
NM_001126045.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500014981.mm.1 |
NM_001126045.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500014981.mm.1 |
NM_001168602.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500014981.mm.1 |
NM_001168602.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500014988.mm.1 |
TC0500003715.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500014988.mm.1 |
NM_001039889.3 |
Refseq |
Mouse Expression Set 430 |
1452516_PM_at |
- |
- |
Mouse Expression Set 430 |
1452516_at |
- |
- |
Murine Genome U74v2 Set |
97007_PM_at |
- |
- |
Murine Genome U74v2 Set |
97007_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1452516_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1452516_PM_at |
- |
- |