Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103171.1 | NP_001096641.1 | 60 - 61 | 10123 | 102 | |
NM_010862.2 | NP_034992.2 | 61 - 62 | 10177 | 102 | ||
NM_182698.2 | NP_874357.2 | 59 - 60 | 6361 | 102 | ||
GenBank mRNA | AF144095.1 | - | 61 - 62 | 10177 | 102 | |
AY331132.1 | - | 60 - 61 | 10123 | 102 | ||
AY331133.1 | - | 59 - 60 | 6361 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |