Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700022789.mm.1 |
TC0700002636.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700022789.mm.1 |
NM_001113368.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022789.mm.1 |
NM_001113368.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022789.mm.1 |
NM_001113369.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022789.mm.1 |
NM_001113369.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022789.mm.1 |
NM_007543.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022789.mm.1 |
NM_007543.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022782.mm.1 |
TC0700002636.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700022782.mm.1 |
NM_001113368.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022782.mm.1 |
NM_001113368.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022782.mm.1 |
NM_001113369.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022782.mm.1 |
NM_001113369.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022788.mm.1 |
TC0700002636.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700022788.mm.1 |
NM_001113368.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022788.mm.1 |
NM_001113368.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022788.mm.1 |
NM_001113369.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022788.mm.1 |
NM_001113369.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022788.mm.1 |
NM_007543.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022788.mm.1 |
NM_007543.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022784.mm.1 |
TC0700002636.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700022784.mm.1 |
NM_001113368.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022784.mm.1 |
NM_001113368.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022784.mm.1 |
NM_001113369.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022784.mm.1 |
NM_001113369.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022784.mm.1 |
NM_007543.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022784.mm.1 |
NM_007543.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022786.mm.1 |
TC0700002636.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700022786.mm.1 |
NM_001113368.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022786.mm.1 |
NM_001113368.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022785.mm.1 |
TC0700002636.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700022785.mm.1 |
NM_001113368.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022785.mm.1 |
NM_001113368.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022791.mm.1 |
TC0700002636.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700022791.mm.1 |
NM_001113368.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022791.mm.1 |
NM_001113368.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022791.mm.1 |
NM_001113369.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022791.mm.1 |
NM_001113369.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022791.mm.1 |
NM_007543.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022791.mm.1 |
NM_007543.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022790.mm.1 |
TC0700002636.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700022790.mm.1 |
NM_001113368.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022790.mm.1 |
NM_001113368.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022790.mm.1 |
NM_001113369.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022790.mm.1 |
NM_001113369.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022790.mm.1 |
NM_007543.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022790.mm.1 |
NM_007543.4 |
Refseq |