Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrgpra3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153067.2 | NP_694707.2 | 1 - 2 | 77 | 80 | |
GenBank mRNA | AY042193.1 | - | 1 - 2 | 92 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mus musculus Probe spans exons |