Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600003137.mm.1 |
TC0600000406.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600003137.mm.1 |
NM_023538.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003137.mm.1 |
NM_023538.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003134.mm.1 |
TC0600000406.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600003134.mm.1 |
NM_023538.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003134.mm.1 |
NM_023538.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003141.mm.1 |
TC0600000406.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600003141.mm.1 |
NM_023538.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003141.mm.1 |
NM_023538.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003142.mm.1 |
TC0600000406.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600003142.mm.1 |
NM_023538.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003142.mm.1 |
NM_023538.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003144.mm.1 |
TC0600000406.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600003144.mm.1 |
NM_023538.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003144.mm.1 |
NM_023538.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003145.mm.1 |
TC0600000406.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600003145.mm.1 |
NM_023538.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003145.mm.1 |
NM_023538.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003148.mm.1 |
TC0600000406.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600003148.mm.1 |
NM_023538.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003148.mm.1 |
NM_023538.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003139.mm.1 |
TC0600000406.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600003139.mm.1 |
NM_023538.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003139.mm.1 |
NM_023538.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003146.mm.1 |
TC0600000406.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600003146.mm.1 |
NM_023538.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003146.mm.1 |
NM_023538.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003147.mm.1 |
TC0600000406.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600003147.mm.1 |
NM_023538.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003147.mm.1 |
NM_023538.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003133.mm.1 |
TC0600000406.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600003133.mm.1 |
NM_023538.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003133.mm.1 |
NM_023538.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003140.mm.1 |
TC0600000406.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600003140.mm.1 |
NM_023538.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003140.mm.1 |
NM_023538.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003149.mm.1 |
TC0600000406.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600003149.mm.1 |
NM_023538.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003149.mm.1 |
NM_023538.2 |
Refseq |