Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.0 ST Array |
10424427 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10424420 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10424423 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10424422 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10424420 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10424421 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10424419 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10424422 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10424421 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10424426 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10424423 |
- |
- |
Affymetrix HT MG-430 PM Array Plate |
1432930_PM_at |
- |
- |
Affymetrix HT MG-430 PM Array Plate |
1432257_PM_at |
- |
- |
Affymetrix HT MG-430 PM Array Plate |
1432929_PM_at |
- |
- |
Affymetrix HT MG-430 PM Array Plate |
1454212_PM_x_at |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10424418 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10424424 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10424418 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10424424 |
- |
- |
Mouse Genome 430 2.0 Array |
1432380_PM_s_at |
- |
- |
Mouse Genome 430 2.0 Array |
1432930_at |
- |
- |
Mouse Genome 430 2.0 Array |
1432257_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1432257_at |
- |
- |
Mouse Genome 430 2.0 Array |
1432930_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1432380_s_at |
- |
- |
Mouse Genome 430 2.0 Array |
1454212_x_at |
- |
- |
Mouse Genome 430 2.0 Array |
1454212_PM_x_at |
- |
- |
Mouse Genome 430 2.0 Array |
1432929_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1432929_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR1500003283.mm.1 |
NR_108053.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003283.mm.1 |
TC1500002332.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500003299.mm.1 |
NR_029414.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003299.mm.1 |
TC1500000469.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500003293.mm.1 |
NR_108051.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003293.mm.1 |
TC1500000468.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500003298.mm.1 |
NR_029414.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003298.mm.1 |
TC1500000469.mm.1 |
MTA TC |
Mouse Expression Set 430 |
1432929_at |
- |
- |
Mouse Expression Set 430 |
1432929_PM_at |
- |
- |
Mouse Expression Set 430 |
1432380_s_at |
- |
- |
Mouse Expression Set 430 |
1432380_PM_s_at |
- |
- |
Mouse Expression Set 430 |
1454212_x_at |
- |
- |
Mouse Expression Set 430 |
1432380_s_at |
- |
- |
Mouse Expression Set 430 |
1432257_at |
- |
- |
Mouse Expression Set 430 |
1432257_PM_at |
- |
- |
Mouse Expression Set 430 |
1432380_PM_s_at |
- |
- |
Mouse Expression Set 430 |
1432930_PM_at |
- |
- |
Mouse Expression Set 430 |
1432930_at |
- |
- |
Mouse Expression Set 430 |
1454212_PM_x_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1432257_PM_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1432257_at |
- |
- |