Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sphk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172472.1 | NP_001165943.1 | 2 - 3 | 449 | 78 | |
NM_001172473.1 | NP_001165944.1 | 2 - 3 | 335 | 78 | ||
NM_025367.6 | NP_079643.2 | 2 - 3 | 332 | 78 | ||
GenBank mRNA | AK155332.1 | - | 2 - 3 | 434 | 78 | |
AK160900.1 | - | 2 - 3 | 325 | 78 | ||
BC037710.1 | - | 2 - 3 | 247 | 4221357 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |