Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.0 ST Array |
10402398 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10402411 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10402400 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10402412 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10402401 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10402400 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10402413 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10402410 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10402411 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10402405 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10402403 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10402401 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10402396 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10402395 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10402397 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10402402 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10402392 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10402403 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10402412 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10402410 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10402391 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10402413 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10402402 |
- |
- |
Affymetrix HT MG-430 PM Array Plate |
1418282_PM_x_at |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10402399 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10402409 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10402390 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10402399 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10402394 |
- |
- |
Mouse Genome 430 2.0 Array |
1418282_PM_x_at |
- |
- |
Mouse Genome 430 2.0 Array |
1451513_PM_x_at |
- |
- |
Mouse Genome 430 2.0 Array |
1449321_PM_x_at |
- |
- |
Mouse Genome 430 2.0 Array |
1449321_x_at |
- |
- |
Mouse Genome 430 2.0 Array |
1418282_x_at |
- |
- |
Mouse Genome 430 2.0 Array |
1451513_x_at |
- |
- |
Affymetrix GeneChip Mouse Genome 430 2.0 Array |
20701_at |
- |
- |
Affymetrix GeneChip Mouse Genome 430 2.0 Array |
20701_PM_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR1200015821.mm.1 |
NM_009244.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015821.mm.1 |
NM_009244.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015821.mm.1 |
TC1200002531.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200018821.mm.1 |
NM_009244.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200018821.mm.1 |
NM_009244.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200018821.mm.1 |
TC1200002531.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200018823.mm.1 |
NM_009246.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200018823.mm.1 |
NM_009246.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200018823.mm.1 |
TC1200002532.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200015819.mm.1 |
NM_009244.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015819.mm.1 |
NM_009244.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015819.mm.1 |
TC1200002531.mm.1 |
MTA TC |
Mouse Expression Set 430 |
1451513_PM_x_at |
- |
- |
Mouse Expression Set 430 |
1449321_x_at |
- |
- |
Mouse Expression Set 430 |
1449321_x_at |
- |
- |
Mouse Expression Set 430 |
1451513_x_at |
- |
- |
Mouse Expression Set 430 |
1418282_x_at |
- |
- |
Mouse Expression Set 430 |
1449321_PM_x_at |
- |
- |
Mouse Expression Set 430 |
1449321_PM_x_at |
- |
- |
Mouse Expression Set 430 |
1418282_PM_x_at |
- |
- |
Murine Genome U74v2 Set |
93109_PM_f_at |
- |
- |
Murine Genome U74v2 Set |
101575_i_at |
- |
- |
Murine Genome U74v2 Set |
93109_f_at |
- |
- |
Murine Genome U74v2 Set |
93109_f_at |
- |
- |
Murine Genome U74v2 Set |
101575_PM_i_at |
- |
- |
Murine Genome U74v2 Set |
101565_PM_f_at |
- |
- |
Murine Genome U74v2 Set |
93109_PM_f_at |
- |
- |
Murine Genome U74v2 Set |
101576_f_at |
- |
- |
Murine Genome U74v2 Set |
101565_f_at |
- |
- |
Murine Genome U74v2 Set |
101565_f_at |
- |
- |
Murine Genome U74v2 Set |
101576_PM_f_at |
- |
- |
Murine Genome U74v2 Set |
101575_PM_i_at |
- |
- |
Murine Genome U74v2 Set |
101565_PM_f_at |
- |
- |
Murine Genome U74v2 Set |
101575_i_at |
- |
- |
Murine Genome U74v2 Set |
101576_PM_f_at |
- |
- |
Murine Genome U74v2 Set |
101576_f_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1449321_PM_x_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1449321_x_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1449321_PM_x_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1418282_PM_x_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1449321_x_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1451513_PM_x_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1418282_x_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1451513_x_at |
- |
- |
Affymetrix HT Mouse Genome 430A Array |
1418282_x_at |
- |
- |
Affymetrix HT Mouse Genome 430A Array |
1418282_PM_x_at |
- |
- |