Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.0 ST Array |
10402412 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10402408 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10402413 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10402401 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10402400 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10402403 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10402407 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10402410 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10402402 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10402411 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10402406 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10402409 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10402399 |
- |
- |
Mouse Genome 430 2.0 Array |
1448680_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1448680_at |
- |
- |
Affymetrix GeneChip Mouse Genome 430 2.0 Array |
20702_at |
- |
- |
Affymetrix GeneChip Mouse Genome 430 2.0 Array |
20702_PM_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR1200015904.mm.1 |
NM_009247.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015904.mm.1 |
NM_009247.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015904.mm.1 |
TC1200002285.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200015844.mm.1 |
NM_001252569.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015844.mm.1 |
NM_001252569.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015844.mm.1 |
NM_009243.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015844.mm.1 |
NM_009243.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015844.mm.1 |
TC1200002533.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200015902.mm.1 |
NM_009247.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015902.mm.1 |
NM_009247.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015902.mm.1 |
TC1200002285.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200015862.mm.1 |
NM_009245.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015862.mm.1 |
NM_009245.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015862.mm.1 |
TC1200002534.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200015863.mm.1 |
NM_009245.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015863.mm.1 |
NM_009245.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015863.mm.1 |
TC1200002534.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200015903.mm.1 |
NM_009247.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015903.mm.1 |
NM_009247.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015903.mm.1 |
TC1200002285.mm.1 |
MTA TC |
Mouse Expression Set 430 |
1449321_x_at |
- |
- |
Mouse Expression Set 430 |
1448680_at |
- |
- |
Mouse Expression Set 430 |
1449321_PM_x_at |
- |
- |
Mouse Expression Set 430 |
1448680_PM_at |
- |
- |
Murine Genome U74v2 Set |
100329_at |
- |
- |
Murine Genome U74v2 Set |
101575_PM_i_at |
- |
- |
Murine Genome U74v2 Set |
101575_i_at |
- |
- |
Murine Genome U74v2 Set |
101576_f_at |
- |
- |
Murine Genome U74v2 Set |
101565_PM_f_at |
- |
- |
Murine Genome U74v2 Set |
93109_PM_f_at |
- |
- |
Murine Genome U74v2 Set |
100329_PM_at |
- |
- |
Murine Genome U74v2 Set |
101565_f_at |
- |
- |
Murine Genome U74v2 Set |
93109_f_at |
- |
- |
Murine Genome U74v2 Set |
101576_PM_f_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1449321_x_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1448680_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1449321_PM_x_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1448680_PM_at |
- |
- |