Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aprt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009698.2 | NP_033828.2 | 3 - 4 | 398 | 103 | |
GenBank mRNA | AB033539.1 | - | 3 - 4 | 381 | 103 | |
AB033540.1 | - | 3 - 4 | 381 | 103 | ||
AK002350.1 | - | 3 - 4 | 351 | 103 | ||
AK153201.1 | - | 3 - 4 | 363 | 103 | ||
BC005667.1 | - | 3 - 4 | 336 | 3152733 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |