Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf2h4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF054823.1 | - | 1 - 2 | 129 | 93 | |
AK079456.1 | - | 1 - 2 | 158 | 93 | ||
AK088296.1 | - | 1 - 2 | 93 | |||
AK144680.1 | - | 1 - 2 | 160 | 93 | ||
AK146112.1 | - | 1 - 2 | 159 | 93 | ||
AK149320.1 | - | 1 - 2 | 160 | 93 | ||
BY109905.1 | - | 1 - 2 | 187 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mus musculus 1700 Probe spans exons |