Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lst1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010734.2 | NP_034864.2 | 1 - 2 | 83 | 89 | |
GenBank mRNA | AI182980.1 | - | 1 - 2 | 83 | 89 | |
BC086881.1 | - | 1 - 2 | 77 | 6747449 | 89 | |
M18187.1 | - | 1 - 2 | 76 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |